Learn More
World Precision Instrument Anesthesia Oxygen Output Splitter
SDP
Catalog No. 502538847
Encompass
  • Y-Splitter for Oxygen Regulator output
  • Allows two EZ Anesthesia Systems to connect to a single Oxygen source
  • For use with EZ Anesthesia systems
Catalog No. 50-253-8847 Supplier World Precision Instrument Supplier No. EZ124DV
  • Y-Splitter for Oxygen Regulator output
  • Allows two EZ Anesthesia Systems to connect to a single Oxygen source
  • For use with EZ Anesthesia systems