Learn More
World Precision Instrument Anesthesia Oxygen Regulator for Large Tank
SDP
Catalog No. 502538821
Encompass
  • For use with EZ Anesthesia systems
  • CGA-540 connection for large 'H' tanks
Regulators are preset to 50 psi.
Catalog No. 50-253-8821 Supplier World Precision Instrument Supplier No. EZ320
  • For use with EZ Anesthesia systems
  • CGA-540 connection for large 'H' tanks
Regulators are preset to 50 psi.