Learn More
World Precision Instrument Anesthesia Oxygen Regulator for Small Tank
SDP
Catalog No. 502538820
Encompass
  • For use with EZ Anesthesia systems
  • CGA-870 connection for small 'E' tanks
Regulators are preset to 50 psi.
Catalog No. 50-253-8820 Supplier World Precision Instrument Supplier No. EZ330
  • For use with EZ Anesthesia systems
  • CGA-870 connection for small 'E' tanks
Regulators are preset to 50 psi.