Learn More
Braintree Scientific Anesthesia Pole Stand w/Mayo Tray
SDP
Catalog No. 501954720
Encompass
Anesthesia Pole Stand for portable transport of EZ Anesthesia systems with a minimal footprint. Accommodates each of our three New Generation EZ Anesthesia systems. Convenient, slide-on mounting. Swivel top allows precise positioning. Accommodates two E-tanks. Stable five-wheel base. Optional Mayo Tray for surgical tools available - call for pricing **Sold for research purposes only and not for human use**
Catalog No. 50-195-4720 Supplier Braintree Scientific Supplier No. E28000PST
Anesthesia Pole Stand for portable transport of EZ Anesthesia systems with a minimal footprint. Accommodates each of our three New Generation EZ Anesthesia systems. Convenient, slide-on mounting. Swivel top allows precise positioning. Accommodates two E-tanks. Stable five-wheel base. Optional Mayo Tray for surgical tools available - call for pricing **Sold for research purposes only and not for human use**