Learn More
Euthanex Anesthesia Pole Stand for E-Z Anesthesia systems with mounting top and five-wheel base with 360 swivel.
Catalog No. 501540144
Encompass_Preferred
Anesthesia Pole Stand, provides for minimal footprint and accommodates each of the three E-Z Systems New Generation E-Z Anesthesia systems. Convenient, slide-on mounting swivel top allows precise positioning ans accommodates two E-tanks. Stable five-wheel base with 360" swivel feature. Optional Mayo Tray for surgical tools is available (E-28000-TRAY). Optional EZ-DOM Pole Mounted Dual Tank Manifold also available. Equipment and tanks not included.
Catalog No. 50-154-0144 Supplier Euthanex Supplier No. E28000
Anesthesia Pole Stand, provides for minimal footprint and accommodates each of the three E-Z Systems New Generation E-Z Anesthesia systems. Convenient, slide-on mounting swivel top allows precise positioning ans accommodates two E-tanks. Stable five-wheel base with 360" swivel feature. Optional Mayo Tray for surgical tools is available (E-28000-TRAY). Optional EZ-DOM Pole Mounted Dual Tank Manifold also available. Equipment and tanks not included.