Learn More
World Precision Instrument Anesthesia/Ventilator Connection Kit
SDP
Catalog No. 502538818
Encompass
Ventilator Connection Kit
This kit is used to connect the CW-SAR-830 ventilators with EZ-Anesthesia Systems. It includes all required components, pre-assembled for simple connection between the ventilator and the anesthesia system, including tubing and fittings.
Catalog No. 50-253-8818 Supplier World Precision Instrument Supplier No. EZ830
Ventilator Connection Kit
This kit is used to connect the CW-SAR-830 ventilators with EZ-Anesthesia Systems. It includes all required components, pre-assembled for simple connection between the ventilator and the anesthesia system, including tubing and fittings.