Description
- The kit includes
- 3 fresh gas supply lines, each 6 feet long
- Common adapters, elbows and 15 mm adapters
- Color-coded for easy identification
- Replace worn or cracked tubing and fittings
- Complete option versus pieces that do not fit
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Animal | Mouse, Rat |
| Type | Connection Kits |
| Disposable | Reusable |
| Size | Standard |
| Sterility | Non-sterile |