Learn More
Medchemexpress LLC Anethole | 104-46-1 | 98.9% | 25 G
SDP
Catalog No. 5000407444
Encompass_Preferred
Anethole is an orally active aromatic compound found widely in nature, primarily used as a flavoring agent. It demonstrates a range of biological effects, including anticancer, anti-inflammatory, antioxidant, and antimicrobial properties. It is also noted for its anesthetic, estrogenic, central nervous system depressant, hypnotic, insecticidal, and gastroprotective activities. This product is for research use only.
  • Exhibits anticancer properties
  • Anti-inflammatory effects
  • Antioxidant activity
  • Antibacterial and antifungal properties
  • Anesthetic and hypnotic effects
  • Central nervous system depressant
  • Gastroprotective properties
  • Used in studies related to oxidative stress-related skin diseases
  • Applicable in prostate cancer research
Catalog No. 50-004-07444 Supplier Medchemexpress LLC Supplier No. 25GHYB0900
Anethole is an orally active aromatic compound found widely in nature, primarily used as a flavoring agent. It demonstrates a range of biological effects, including anticancer, anti-inflammatory, antioxidant, and antimicrobial properties. It is also noted for its anesthetic, estrogenic, central nervous system depressant, hypnotic, insecticidal, and gastroprotective activities. This product is for research use only.
  • Exhibits anticancer properties
  • Anti-inflammatory effects
  • Antioxidant activity
  • Antibacterial and antifungal properties
  • Anesthetic and hypnotic effects
  • Central nervous system depressant
  • Gastroprotective properties
  • Used in studies related to oxidative stress-related skin diseases
  • Applicable in prostate cancer research