Learn More
Medchemexpress LLC Anethole | 104-46-1 | 99.8% | 100 G
SDP
Catalog No. 5000409347
Encompass_Preferred
Anethole is a naturally occurring aromatic compound widely used as a flavoring agent. It exhibits various biological activities, such as anticancer, anti-inflammatory, antioxidant, antibacterial, and antifungal effects. This compound also demonstrates anesthetic, estrogenic, central nervous system depressant, hypnotic, insecticidal, and gastroprotective properties, useful for research into oxidative stress-related skin diseases and prostate cancer.
  • Orally active aromatic compound
  • Widely found in nature
  • Used as a flavoring agent
  • Exhibits anticancer, anti-inflammatory, and antioxidant effects
  • Shows antibacterial, antifungal, and anesthetic properties
  • Demonstrates estrogenic, central nervous system depressant, and hypnotic effects
  • Possesses insecticidal and gastroprotective benefits
  • Useful in research for oxidative stress-related skin diseases and prostate cancer
Catalog No. 50-004-09347 Supplier Medchemexpress LLC Supplier No. HYB0900100G
Anethole is a naturally occurring aromatic compound widely used as a flavoring agent. It exhibits various biological activities, such as anticancer, anti-inflammatory, antioxidant, antibacterial, and antifungal effects. This compound also demonstrates anesthetic, estrogenic, central nervous system depressant, hypnotic, insecticidal, and gastroprotective properties, useful for research into oxidative stress-related skin diseases and prostate cancer.
  • Orally active aromatic compound
  • Widely found in nature
  • Used as a flavoring agent
  • Exhibits anticancer, anti-inflammatory, and antioxidant effects
  • Shows antibacterial, antifungal, and anesthetic properties
  • Demonstrates estrogenic, central nervous system depressant, and hypnotic effects
  • Possesses insecticidal and gastroprotective benefits
  • Useful in research for oxidative stress-related skin diseases and prostate cancer