Learn More
Surgical Design™ Aneurysm Needle Blunt Tip With Eye
Catalog No. 21124621 Shop All Surgical Design Products
Change view
Click to view available options
Length (English):
6.5 in.
Catalog No. Length (English)
21-124-621 6.5 in.
1 options
Catalog No. 21-124-621 Supplier Surgical Design™ Supplier No. ATPM001
Add to Cart
Add to Cart
Aneurysm needle, 6.5 in., with blunt tip and eyelet supports controlled vessel looping, tissue separation, and anatomical exploration in postmortem exams.
  • Made from high-grade stainless steel, built to withstand repeated use in autopsy and pathology labs
  • Blunt, rounded tip ensures safe manipulation and separation of delicate vessels and soft tissues without perforation
  • Eyelet opening accommodates twine, ligature, or vessel loops for securing and isolating arteries during dissection
  • 6.5-in. length provides optimal reach for thoracic, abdominal, cervical, and neurovascular examinations
  • Long, narrow shaft ideal for tracing vessels, exposing anatomical pathways, and elevating tissues during gross dissection
  • Ergonomic grip offers stability and comfort during extended autopsy procedures
  • Corrosion-resistant satin finish minimizes glare under strong pathology lab lighting
  • Fully autoclavable and reusable, meeting mortuary and forensic laboratory sterilization needs
  • Vessel isolation during organ removal
  • Tracing arterial pathways in cardiovascular evaluation
  • Manipulating soft tissues around the neck, thorax, and abdomen
  • Useful in forensic, pathology, mortuary, and educational anatomy settings
Specifications
Product Type Needle Probe
Autoclavable Autoclavable
Certifications/Compliance ISO/GMP
Color Silver
Material Stainless Steel
Latex Free Yes
Length (English) 6.5 in.
Sterility Non-sterile