Learn More
MSC Vikan Washing Brush, Angle Adjustment, Waterfed, 240 mm, Soft/Split, Blue
Catalog No. 502836170
Encompass
This fully adjustable, waterfed washing brush efficiently removes dust and dirt from high, difficult-to-reach areas. It features split fiber bristles that retain water and cleaning chemicals, enhancing cleaning efficacy, and is compatible with various handles.
  • For food manufacturing, retail, and service, ensuring hygiene and safety.
  • Soft/split bristles prevent surface damage and retain water/chemicals for maximum washing.
  • Adjustable angle for cleaning walls, windows, tanks, and high surfaces.
  • Compatible with long or short handles.
  • Safe for sensitive surfaces.
  • Waterfed design for wet cleaning.
  • Even water distribution.
  • Easy to clean and maintain for hygiene.
  • Durable construction for long-lasting performance.
  • Colour-coded for hygienic zoning and 5S programs.
  • Compatible with Euro thread and waterfed handles.
Catalog No. 50-283-6170 Supplier MSC Supplier No. 70247580
This fully adjustable, waterfed washing brush efficiently removes dust and dirt from high, difficult-to-reach areas. It features split fiber bristles that retain water and cleaning chemicals, enhancing cleaning efficacy, and is compatible with various handles.
  • For food manufacturing, retail, and service, ensuring hygiene and safety.
  • Soft/split bristles prevent surface damage and retain water/chemicals for maximum washing.
  • Adjustable angle for cleaning walls, windows, tanks, and high surfaces.
  • Compatible with long or short handles.
  • Safe for sensitive surfaces.
  • Waterfed design for wet cleaning.
  • Even water distribution.
  • Easy to clean and maintain for hygiene.
  • Durable construction for long-lasting performance.
  • Colour-coded for hygienic zoning and 5S programs.
  • Compatible with Euro thread and waterfed handles.