Learn More
American Radiolabeled Chemicals Inc ANG I TYR4 125I 10UC
SDP
Catalog No. 50901173
Encompass
Angiotensin I, Tyr4-125I (human) 10uCi in tris buffer, 0.05 mCi/ml, S.A. 2200 Ci/mmol, M.W. ~1420
Catalog No. 50-901-173 Supplier American Radiolabeled Chemicals Inc Supplier No. ARI 016910UC
Angiotensin I, Tyr4-125I (human) 10uCi in tris buffer, 0.05 mCi/ml, S.A. 2200 Ci/mmol, M.W. ~1420