Learn More
American Radiolabeled Chemicals Inc ANG II TYR4 125I 50UC
SDP
Catalog No. 50901072
Encompass
Angiotensin II, Tyr4-125I (human) 50uCi in tris buffer, 0.05 mCi/ml, S.A. 2200 Ci/mmol, M.W. ~1171
Catalog No. 50-901-072 Supplier American Radiolabeled Chemicals Inc Supplier No. ARI 017050UC
Add to Cart
Add to Cart
Request a Quote
Angiotensin II, Tyr4-125I (human) 50uCi in tris buffer, 0.05 mCi/ml, S.A. 2200 Ci/mmol, M.W. ~1171