Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Angelic acid | 565-63-9 | MFCD00002654 | 5 MG
SDP

Catalog No. 5000420119
Encompass_Preferred

Angelic acid is a ferroptosis inducer that targets NRF2 degradation. It binds to the NRF2 protein, promoting its degradation through the ubiquitination-proteasome pathway. This action mitigates the inhibitory effect of NRF2 on oxidative stress and lipid peroxidation, leading to the induction of ferroptosis in tumor cells.

  • Purity: 99.20%
  • Molecular weight: 100.12
  • Molecular formula: C5H8O2
  • Appears as a white to off-white solid
  • Enhances accumulation of intracellular reactive oxygen species
  • Upregulates ferroptosis-related markers such as CHAC1 and PTGS2
  • Synergizes with other ferroptosis inducers to boost anti-tumor effects
  • Scavenges UVA-induced reactive oxygen species
  • Inhibits skin fibroblast senescence and prevents extracellular matrix degradation
  • Contributes to wound healing and has sedative activity

Catalog No. 50-004-20119 Supplier Medchemexpress LLC Supplier No. HYN69295MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Angelic acid is a ferroptosis inducer that targets NRF2 degradation. It binds to the NRF2 protein, promoting its degradation through the ubiquitination-proteasome pathway. This action mitigates the inhibitory effect of NRF2 on oxidative stress and lipid peroxidation, leading to the induction of ferroptosis in tumor cells.

  • Purity: 99.20%
  • Molecular weight: 100.12
  • Molecular formula: C5H8O2
  • Appears as a white to off-white solid
  • Enhances accumulation of intracellular reactive oxygen species
  • Upregulates ferroptosis-related markers such as CHAC1 and PTGS2
  • Synergizes with other ferroptosis inducers to boost anti-tumor effects
  • Scavenges UVA-induced reactive oxygen species
  • Inhibits skin fibroblast senescence and prevents extracellular matrix degradation
  • Contributes to wound healing and has sedative activity

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.