Learn More
Selleck Chemical LLC Angelic anhydride S3806-25mg
SDP
Catalog No. 503109012
Encompass_Preferred
Angelic anhydride (2-Methylisocrotonic Anhydride (Z)-2-Methylbut-2-enoic anhydride) is a synthetic chemical and usually used in content determination assays and pharmacological experiments
Catalog No. 50-310-9012 Supplier Selleck Chemical LLC Supplier No. S380625MG
Add to Cart
Add to Cart
Angelic anhydride (2-Methylisocrotonic Anhydride (Z)-2-Methylbut-2-enoic anhydride) is a synthetic chemical and usually used in content determination assays and pharmacological experiments