Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Angelic Anhydride 98.0+%, TCI America™
SDP

Catalog No. A11805G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A11805G 5 g
A11801G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A11805G Supplier TCI America Supplier No. A11805G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 94487-74-8
Molecular Formula C10H14O3
Molecular Weight (g/mol) 182.219
MDL Number MFCD00198080
InChI Key LIHLSLRANKCHLV-SFECMWDFSA-N
Synonym (Z)-2-Methyl-2-butenoic Anhydride, 2-Methylisocrotonic Anhydride
PubChem CID 44630058
IUPAC Name [(Z)-2-methylbut-2-enoyl] (Z)-2-methylbut-2-enoate
SMILES CC=C(C)C(=O)OC(=O)C(=CC)C

Specifications

Color Yellow
Boiling Point 108°C
Quantity 5 g
UN Number 3265
Formula Weight 182.22
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material Angelic Anhydride
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.