Learn More
Medchemexpress LLC α-Angelica lactone | 591-12-8 | 98.10 | 500 MG
SDP
Supplier:  Medchemexpress LLC HYN0548500MG
Encompass_Preferred
α-Angelica lactone is a naturally occurring anticarcinogen and a vinylogous nucleophile. It can produce chiral δ-amino γ,γ-disubstituted butenolide carbonyl derivatives and demonstrates electrophilic trapping at the γ-carbon. This compound exhibits strong chemoprotective effects by selectively enhancing glutathione-S-thansferase (GST) and UDP-glucononosyltransferase (UGT) detoxification enzymes.
  • Naturally occurring anticarcinogen
  • Exhibits strong chemoprotective effects
  • Enhances glutathione-S-thansferase (GST) detoxification enzyme activity
  • Enhances UDP-glucononosyltransferase (UGT) detoxification enzyme activity
Catalog No. 50-003-88565