Learn More
Medchemexpress LLC Angeloylgomisin Q | 72561-28-5 | 1 MG
SDP
Catalog No. 5000417477
Encompass_Preferred
Angeloylgomisin Q is a new dibenzocyclooctadiene lignan isolated from the stems of *Schisandra sphaerandra*. It has potential for Alzheimer's disease research.
  • New dibenzocyclooctadiene lignan
  • Isolated from *Schisandra sphaerandra* stems
  • Potential for Alzheimer's disease research
Catalog No. 50-004-17477 Supplier Medchemexpress LLC Supplier No. HYN82601MG
Angeloylgomisin Q is a new dibenzocyclooctadiene lignan isolated from the stems of *Schisandra sphaerandra*. It has potential for Alzheimer's disease research.
  • New dibenzocyclooctadiene lignan
  • Isolated from *Schisandra sphaerandra* stems
  • Potential for Alzheimer's disease research