Learn More
Medchemexpress LLC HY-P2341 5mg , Angiopep-2 (hydrochloride) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP23415MG
Encompass_Preferred
HY-P2341 5mg Angiopep-2 (hydrochloride) CAS: Angiopep-2 hydrochloride is a brain peptide vector. The conjugation of anticancer agents with the Angiopep-2 peptide vector could increase their efficacy in the treatment of brain cancer. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1211