Learn More
Medchemexpress LLC Angiopeptin TFA | 2478421-60-0 | 99.01% | 1326.39 | 25 MG
SDP
Catalog No. 5000426918
Encompass_Preferred
Angiopeptin TFA is a cyclic octapeptide analogue of somatostatin. It acts as a weak sst2/sst5 receptor partial agonist with IC50 values of 0.26 nM and 6.92 nM, respectively. It is a potent inhibitor of growth hormone release and insulin-like growth factor-1 (IGF-1) production. Angiopeptin TFA also inhibits adenylate cyclase or stimulates extracellular acidification, making it valuable for research in coronary atherosclerosis.
  • Acts as a partial agonist on the release of tritium on CHO hsst2 cells (in vitro).
  • Significantly inhibits neointimal formation (in vivo).
  • Significantly inhibits coronary artery myointimal proliferation in cardiac allografts (in vivo).
Catalog No. 50-004-26918 Supplier Medchemexpress LLC Supplier No. 25MGHYP2090A
Angiopeptin TFA is a cyclic octapeptide analogue of somatostatin. It acts as a weak sst2/sst5 receptor partial agonist with IC50 values of 0.26 nM and 6.92 nM, respectively. It is a potent inhibitor of growth hormone release and insulin-like growth factor-1 (IGF-1) production. Angiopeptin TFA also inhibits adenylate cyclase or stimulates extracellular acidification, making it valuable for research in coronary atherosclerosis.
  • Acts as a partial agonist on the release of tritium on CHO hsst2 cells (in vitro).
  • Significantly inhibits neointimal formation (in vivo).
  • Significantly inhibits coronary artery myointimal proliferation in cardiac allografts (in vivo).