Learn More
Enzo Life Sciences COMP (rat) : Angiopoietin 1 (human), (recombinant) (500µg)
SDP
Supplier:  Enzo Life Sciences ALX201314C500
Encompass_Preferred
Soluble, stable and more potent Ang-1 variant. Soluble, stable and more potent Ang-1 variant. Alternative name: COMP:Ang-1. Concentration: ~0.2-0.4 mg/mL (Please refer to Certificate of Analysis for lot-specific concentration.). Endotoxin: <0.1EU/µg protein (LAL test; BioWhittaker). Purity: ≥90% (SDS-PAGE). Formulation: Liquid. 0.2µm-filtered solution in PBS containing 0.01% CHAPS. Source: Produced in CHO cells. The fibrinogen-like domain (aa 284-498) of human Ang-1 (angiopoietin-1) is fused at the N-terminus to the coiled-coil domain of rat COMP (cartilage oligomeric matrix protein) and to a FLAG-tag. Specificity: Binds to human Tie-2. Long Term Storage: -80°C. Use/Stability: Working aliquots are stable for up to 3 months when stored at -80°C. Handling: Avoid freeze/thaw cycles. After opening, prepare aliquots and store at -80°C.
Catalog No. 50-200-7973
Add to Cart
Request a Quote