Learn More
Sino Biological Human Angiopoietin 1 / ANG1 / ANGPT1 Protein (Fc Tag) 100ug
Supplier:  Sino Biological 13667H02H1100
Encompass_Preferred
A DNA sequence encoding the human ANGPT1 ( NP_001137.2) (Met1-Leu261 and Arg277-Phe498 linked by a polypeptide linker) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-211-4975