Learn More
Medchemexpress LLC Angiopoietin-4 Huma 100ug
SDP
Catalog No. 5000178895
Encompass_Preferred
Angiopoietin 4 protein (ANGPT4) critically binds to TEK/TIE2 regulates ANGPT1 signaling and induces TEK/TIE2 phosphorylation In addition to regulation ANGPT4 promotes endothelial cell survival migration and angiogenesis Angiopoietin-4 Protein Human (HEK293 Fc) is the recombinant human-derived Angiopoietin-4 protein expressed by HEK293 with N-hFc labeled tag
Catalog No. 50-001-78895 Supplier Medchemexpress LLC Supplier No. HYP74411100UG
Add to Cart
Add to Cart
Angiopoietin 4 protein (ANGPT4) critically binds to TEK/TIE2 regulates ANGPT1 signaling and induces TEK/TIE2 phosphorylation In addition to regulation ANGPT4 promotes endothelial cell survival migration and angiogenesis Angiopoietin-4 Protein Human (HEK293 Fc) is the recombinant human-derived Angiopoietin-4 protein expressed by HEK293 with N-hFc labeled tag