Learn More
ACROBiosystems Angiopoietin-like 3 100ug
Catalog No. 502535507
Encompass_Preferred
Human Angiopoietin-like 3 Protein Fc Tag (AN3-H5253) is expressed from human 293 cells (HEK293). It contains AA Ser 17 - Glu 460 (Accession # Q9Y5C1-1).
Catalog No. 50-253-5507 Supplier ACROBiosystems Supplier No. AN3H5253100UG
Human Angiopoietin-like 3 Protein Fc Tag (AN3-H5253) is expressed from human 293 cells (HEK293). It contains AA Ser 17 - Glu 460 (Accession # Q9Y5C1-1).