Learn More
Medchemexpress LLC HY-P7508 10ug Medchemexpress, Angiopoietin-Related Protein 4, C-Fc (HEK293-expressed), Mouse CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP750810UG
Encompass_Preferred
Medchemexpress, HY-P7508 10ug Angiopoietin-Related Protein 4, C-Fc (HEK293-expressed), Mouse CAS: Recombinant Mouse Angiopoietin-Related Protein 4, C-Fc (HEK293-expressed) expresses in HEK293 with an Fc fragment at the C-terminus. Angiopoietin-Related Protein 4 (ANGPTL4) is a multifunctional cytokine regulating vascular permeability, angiogenesis, and inflammation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3172