Learn More
ACROBiosystems Recombinant Protein;Human Angiopoietin-2 / ANGPT2 (275-496) Protein;HEK293;500UG;AN2,ANGPT2,ANGPT-2,AGPT2,AGPT-2,ANG2,ANG-2,Angiopoietin-2,Angiopoietin 2
Catalog No. 501659061
Encompass_Preferred
Recombinant Protein;Human Angiopoietin-2 / ANGPT2 (275-496) Protein;500UG;Human ANGPT2 (275-496), His Tag (AN2-H5242) is expressed from human 293 cells (HEK293). It contains AA Lys 275 - Phe 496 (Accession # AAI26201.1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 26.3 kDa. The protein migrates as 30-33 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-9061 Supplier ACROBiosystems Supplier No. AN2H5242500UG
Recombinant Protein;Human Angiopoietin-2 / ANGPT2 (275-496) Protein;500UG;Human ANGPT2 (275-496), His Tag (AN2-H5242) is expressed from human 293 cells (HEK293). It contains AA Lys 275 - Phe 496 (Accession # AAI26201.1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 26.3 kDa. The protein migrates as 30-33 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.