Learn More
ACROBiosystems Recombinant Protein;Human Angiopoietin-like 3 / ANGPTL3 Protein;HEK293;1MG;AN3,ANGPTL3,ANGPT5,ANG-5,Angiopoietin-5,ANGPTL-3,ANGPT-5,ANG5,Angiopoietin 5,FHBL2,FHBL-2
Catalog No. 501659059
Encompass_Preferred
Recombinant Protein;Human Angiopoietin-like 3 / ANGPTL3 Protein;1MG;Human ANGPTL3, His Tag (AN3-H5226) is expressed from human 293 cells (HEK293). It contains AA Ser 17 - Pro 220 (Accession # NP_055310).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 24.6 kDa. The protein migrates as 26-38 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-9059 Supplier ACROBiosystems Supplier No. AN3H52261MG
Recombinant Protein;Human Angiopoietin-like 3 / ANGPTL3 Protein;1MG;Human ANGPTL3, His Tag (AN3-H5226) is expressed from human 293 cells (HEK293). It contains AA Ser 17 - Pro 220 (Accession # NP_055310).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 24.6 kDa. The protein migrates as 26-38 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.