Learn More
3B Scientific™ Angiospermae I. Gymnospermae Slide Set
SDPSCI_ED
Catalog No. S95054L Shop All 3B Scientific Products
Change view
Click to view available options
Quantity:
15
Catalog No. Quantity
S95054L 15
1 options
Catalog No. S95054L Supplier 3B Scientific™ Supplier No. W13045
Add to Cart
Add to Cart
15 microscope slides
  • 1(e). Ephedra, male cone l.s.
  • 2(f). Ephedra, female cone at pollination time l.s.
  • 3(c). Ginkgo, young sprout, t.s.
  • 4(c). Ginkgo, leaf t.s.
  • 5(c). Pinus, pine, young root
  • 6(c). Pinus, pine, first year stem
  • 7(e). Pinus, pine, bud showing vascular anatomy and origin of leaves l.s.
  • 8(d). Pinus, pine, wood, transverse, radial and tangential sections
  • 9(c). Pinus, pine, needles (leaves) t.s.
  • 10(b). Pinus, pine, w.m. of mature pollen grains
  • 11(d). Pinus, pine, male cone l.s.
  • 12(d). Pinus, pine, young female cone l.s.
  • 13(c). Larix, larch, t.s. of needles (leaves) t.s.
  • 14(d). Larix, larch, male cone l.s.
  • 15(e). Larix, larch, female cone with ovules l.s.
Specifications
Type Gymnospermae
Quantity 15