Learn More
3B Scientific™ Angiospermae Roots Slide Set
SDPSCI_ED
Catalog No. S95056L Shop All 3B Scientific Products
Change view
Click to view available options
Quantity:
15
Catalog No. Quantity
S95056L 15
1 options
Catalog No. S95056L Supplier 3B Scientific™ Supplier No. W13047
Add to Cart
Add to Cart
15 microscope slides
  • 1(d). Allium cepa, onion, root tips, l.s. showing all stages of mitosis
  • 2(c). Zea mays, corn, t.s. of typical monocot root
  • 3(c). Iris, t.s. of typical monocot root
  • 4(c). Ranunculus, buttercup, t.s. of a typical dicot root
  • 5(c). Sarothamnus, broom, t.s. through woody root
  • 6(c). Taraxacum, dandelion, t.s. through tap root showing lactiferous ducts
  • 7(d). Vicia faba, bean, root nodule t.s. nitrogen fixing bacteria
  • 8(d). Ranunculus ficaria, tuber during fall season, t.s. showing starch
  • 9(d). Alnus, alder, t.s. of tuber showing actinomycetes
  • 10(d). Neottia, orchid, t.s. of root with endotrophic mycorrhiza
  • 11(d). Cuscuta, dodder, on host, t.s. haustorium
  • 12(d). Root hairs, w.m. of root tip, root cap and root hairs
  • 13(d). Zea mays, root tip, median l.s. showing central pith, cap and starch
  • 14(c). Monstera, aerial root t.s.
  • 15(c). Elodea, Canadian waterweed, t.s. of an aquatic root
Specifications
Type Angiospermae Roots
Quantity 15