Learn More
3B Scientific™ Angiospermae Stems Slide Set
SDPSCI_ED
Catalog No. S95057L Shop All 3B Scientific Products
Change view
Click to view available options
Quantity:
20
Catalog No. Quantity
S95057L 20
1 options
Catalog No. S95057L Supplier 3B Scientific™ Supplier No. W13048
Add to Cart
Add to Cart
20 microscope slides
  • 1(c). Canna, t.s. of typical monocot stem with scattered bundles
  • 2(f). Aristolochia, t.s. of one year, two years stem and older stem, all 3 in on slide
  • 3(e). Dicot and monocot stem, t.s. of Helianthus and Canna
  • 4(e). Dicot and monocot stem, t.s. of Ranunculus and Zea
  • 5(e). Tilia, lime, two t.s. of stems, first year and two years
  • 6(d). Fagus silvatica, beech, three sections of wood, t.s., r.l.s., t.l.s.
  • 7(d). Fraxinus excelsior, ash, three sections of wood, t.s., r.l.s., t.l.s.
  • 8(c). Quercus, oak, t.s. of stem showing cambium and bark
  • 9(c). Sambucus, elder, t.s. of bark showing lenticells
  • 10(c). Linum, flax, t.s. of stem showing husk fibres
  • 11(b). Linum, flax, isolated husk fibres, w.m.
  • 12(d). Ranunculus, l.s. of herbaceous stem
  • 13(d). Cucurbita pepo, l.s. of stem with sieve tubes
  • 14(d). Sieve plates in top view, t.s. of Cucurbita stem
  • 15(c). Lamium, t.s. of square stem, collenchyma
  • 16(c). Secale, rye, t.s. of typical grass stem
  • 17(c). Nymphaea, water lily, t.s. of aquatic stem, spicular cells
  • 18(c). Hippuris, t.s. of typical aquatic stem with large central pith
  • 19(d). Urtica, nettle, stinging hairs with poison ducts
  • 20(c). Solanum tuberosum, potato, t.s. of tuber with starch grains and cork
Specifications
Type angiospermae stems
Quantity 20