Learn More
Apexbio Technology LLC Angiotensin (1-7), 10mg. Cas: 51833-78-4 MFCD: MFCD00153513. Vasoconstriction peptide hormone
SDP
Supplier:  Apexbio Technology LLC A104110
Encompass_Preferred
Ang-(1-7) (H - Asp - Arg - Val - Tyr - Ile - His - Pro - OH) is an endogenous peptide fragment that can be produced from Ang I or Ang II via endo- or carboxy-peptidases respectively[1]. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4676