Learn More
Medchemexpress LLC HY-12403 5mg Medchemexpress, Angiotensin (1-7) CAS:51833-78-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY124035MG
Encompass_Preferred
Medchemexpress, HY-12403 5mg Angiotensin (1-7) CAS:51833-78-4 Angiotensin 1-7 (Ang-(1-7)) is a major active component of the renin-angiotensin system (RAS), produced from cleavage of Ang II by angiotensin-converting-enzyme type 2 (ACE2). Angiotensin 1-7 inhibits purified canine ACE activity (IC50=0.65 μM). Angiotensin 1-7 acts as a local synergistic modulator of kinin-induced vasodilation by inhibiting ACE and releasing nitric oxide. Angiotensin 1-7 blocks Ang II-induced smooth muscle cell proliferation and hypertrophy and shows antiangiogenic and growth-inhibitory effects on the endothelium. Angiotensin 1-7 shows anti-inflammatory activity [2][3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9976