Learn More
Biomatik Corporation Angiotensin 1-7 (Ang1-7) Monoclonal Antibody
Supplier:  Biomatik Corporation CAU29564
Encompass_Preferred
Angiotensin 1-7 (Ang1-7) Monoclonal Antibody is a Human (Homo sapiens) Monoclonal Antibody to Angiotensin 1-7 (Ang1-7). The Angiotensin 1-7 (Ang1-7) Monoclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Mouse. Species: Human (Homo sapiens). Immunogen: Asp34~Pro40. Applications: WB, IHC, ICC, IP. Target Synonyms: Angiotensin 1-7 (Ang1-7). Accession Number: P01019. Gene ID: 183. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5400