Learn More
Bioworld ANGIOTENSIN
Supplier:  Bioworld 220602351
Encompass_Preferred
Angiotensin Description: Angiotensin is an oligopeptide in the blood that causes vasoconstriction, increased blood pressure, and release of aldosterone from the adrenal cortex. It is a powerful dipsogen. It is derived from the precursor...
Catalog No. 50-253-335