Learn More
Sigma Aldrich Fine Chemicals Biosciences Angiotensin Converting Enz
Supplier:  Sigma Aldrich Fine Chemicals Biosciences A2580.5UN
Encompass
Angiotensin converting enzyme (ACE) is encoded by the gene mRNA-decapping enzyme subunit 1 (DCP1). It is a zinc metalloprotease which belongs to the M2 family.
Catalog No. 50-290-7389