Learn More
CPC Scientific Glp-Trp-Pro-Arg-Pro-Gln-Ile-Pro-Pro-OH (trifluoroacetate salt) 1MG
SDP
Catalog No. 502108573
Encompass
Nonapeptide capable of inhibiting the conversion of angiotensin I to angiotensin II, in vitro and in vivo. Bradykinin Potentiator.SEQUENCE: Glp-Trp-Pro-Arg-Pro-Gln-Ile-Pro-Pro-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: Glp-WPRPQIPPMOLECULAR FORMULA: C53H76N14O12MOLECULAR WEIGHT: 1101.3STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [35115-60-7]SYNONYMS: RESEARCH AREA: CardiovascularREFERENCES:M.A. Ondetti et al., Biochemistry, 10, 4033 (1971)H.S. Cheung and D.W. Cushman, Biochim. Biophys. Acta, 293, 451 (1973)L.C. Martin et al., Biochem. J., 184, 713 (1979)D.I. Marlborough et al., Arch. Biochem. Biophys., 210, 43 (1981)SKU(s): ACEI-001A, ACEI-001B, ACEI-001C
Catalog No. 50-210-8573 Supplier CPC Scientific Supplier No. ACEI001A
Nonapeptide capable of inhibiting the conversion of angiotensin I to angiotensin II, in vitro and in vivo. Bradykinin Potentiator.SEQUENCE: Glp-Trp-Pro-Arg-Pro-Gln-Ile-Pro-Pro-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: Glp-WPRPQIPPMOLECULAR FORMULA: C53H76N14O12MOLECULAR WEIGHT: 1101.3STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [35115-60-7]SYNONYMS: RESEARCH AREA: CardiovascularREFERENCES:M.A. Ondetti et al., Biochemistry, 10, 4033 (1971)H.S. Cheung and D.W. Cushman, Biochim. Biophys. Acta, 293, 451 (1973)L.C. Martin et al., Biochem. J., 184, 713 (1979)D.I. Marlborough et al., Arch. Biochem. Biophys., 210, 43 (1981)SKU(s): ACEI-001A, ACEI-001B, ACEI-001C