Learn More
Medchemexpress LLC Angiotensin I-13C6,15N (human, mouse, rat) TFA | 484-42-4 | 99.8% | 1303.42 (free base) | 1 MG
SDP
Catalog No. 5000426274
Encompass_Preferred
Angiotensin I-13C6,15N (human, mouse, rat) TFA is a 13C and 15N labeled compound. It is a precursor of angiotensin II and is cleaved into angiotensin II by angiotensin converting enzyme (ACE). This product is for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-26274 Supplier Medchemexpress LLC Supplier No. HYP1032S21MG
Angiotensin I-13C6,15N (human, mouse, rat) TFA is a 13C and 15N labeled compound. It is a precursor of angiotensin II and is cleaved into angiotensin II by angiotensin converting enzyme (ACE). This product is for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS