Learn More
Enzo Life Sciences Angiotensin I (5mg)
SDP
Supplier:  Enzo Life Sciences ALX151037M005
Encompass_Preferred
Formula: C62H89N17O14. MW: 1296.5. Purity: ≥97% (HPLC). Formulation: TFA salt. Solubility: Soluble in water. Long Term Storage: -20°C.
Catalog No. 50-200-7772