Learn More
CPC Scientific Glp-Gly-Leu-Pro-Pro-Gly-Pro-Pro-Ile-Pro-Pro-OH (trifluoroacetate salt) 5MG
SDP
Supplier:  CPC Scientific ACEI003B
Encompass
Angiotensin I-converting enzyme inhibitor. Bradykinin Potentiator C.ONE-LETTER SEQUENCE: Glp-GLPPGPPIPPMOLECULAR FORMULA: C51H77N11O13MOLECULAR WEIGHT:1052.3STORAGE CONDITIONS: -20 5CRESEARCH AREA: CardiovascularREFERENCES: H. Kato et al., Biochemistry, 10, 972 (1971)
Catalog No. 50-210-8579