Learn More
CPC Scientific Glp-Gly-Leu-Pro-Pro-Arg-Pro-Lys-Ile-Pro-Pro-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific ACEI002A
Encompass
Angiotensin I-converting enzyme inhibitor. Bradykinin Potentiator B.ONE-LETTER SEQUENCE: Glp-GLPPRPKIPPMOLECULAR FORMULA: C56H91N15O13MOLECULAR WEIGHT:1182.4STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [30892-86-5], RESEARCH AREA: CardiovascularREFERENCES: H. Kato et al., Biochemistry, 10, 972 (1971)
Catalog No. 50-210-8576