Learn More
Bachem Angiotensin I/II (3-8)
SDP
Supplier:  Bachem 4018068.0025
Encompass_Preferred
Angiotensin I/II (3-8), H-Val-Tyr-Ile-His-Pro-Phe-OH, 23025-68-5, C40H54N8O8, Mr 774.92, 25mg. Angiotensin IV, Angiotensin II (3-8), human. Ang IV, one of the major metabolites of angiotensin II, binds with high affinity to a new angiotensin binding site distinct and separate from angiotensin II receptors.
Catalog No. 50-260-178