Learn More
Bachem Angiotensin II Acetate
SDP
Supplier:  Bachem 4006473.0100
Encompass_Preferred
Angiotensin II Acetate, H-Asp-Arg-Val-Tyr-Ile-His-Pro-Phe-OH, 4474-91-3, C50H71N13O12, Mr 1046.19, 100mg. Ang II, Angiotensin II, human. Angiotensin II, DRVYIHPF, is the major biologically active component of the renin-angiotensin system. Its activities include the regulation of blood pressure, salt and water balance, and vascular tonus. Ang II stimulates various enzymes such as NADPH oxidase. Ang II is often used as a model compound in the development and evaluation of analytical methods for peptides and proteins (e.g. MS).
Catalog No. 50-259-527