Learn More
Enzo Life Sciences Angiotensin II (25mg). CAS: 4474-91-3
SDP
Supplier:  Enzo Life Sciences ALX151039M025
Encompass_Preferred
Formula: C50H71N13O12. MW: 1046.2. Purity: ≥97% (HPLC). Appearance: Lyophilized. Formulation: Acetate salt. Solubility: Soluble in water. Long Term Storage: -20°C.
Catalog No. 50-200-7774