Learn More
Apexbio Technology LLC Angiotensin II, 5mg. Cas: 4474-91-3 MFCD: MFCD00161484. Potent vasopressor and a powerful stimulus for production and release of aldosterone from the adrenal cortex.
SDP
Supplier:  Apexbio Technology LLC A10425
Encompass_Preferred
Angiotensin II (Asp-Arg-Val-Tyr-Ile-His-Pro-Phe | His-Leu) is an octapeptide that is converted from Angiotensin I (AI) through removal of two C-terminal residues by the enzyme angiotensin-converting enzyme (ACE), primarily through ACE within the lung. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4680
 Restricted Item