Learn More
CPC Scientific H-Asp-Arg-Val-Tyr-Ile-His-Pro-Phe-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific ANGT003A
Encompass
SEQUENCE: H-Asp-Arg-Val-Tyr-Ile-His-Pro-Phe-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: DRVYIHPFMOLECULAR FORMULA: C50H71N13O12MOLECULAR WEIGHT: 1046.20STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [4474-91-3]SYNONYMS: Angiotensin 2, Ang IIRESEARCH AREA: CardiovascularREFERENCES:A.W.A. Hahn et al., J. Cardiovasc. Pharmacol., 22 (Suppl.5), S37 (1993)T. Inagami et al., Cardiovase. Drugs Ther., 2, 453 (1988)SKU(s): ANGT-003A
Catalog No. 50-210-8764