Learn More
TARGETMOL CHEMICALS INC ANGIOTENSIN II HUMAN 25MG
SDP
Catalog No. 502576361
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Angiotensin II is mediated by AT1 and AT2 receptors. AT1 and AT2 receptors have seven transmembrane glycoproteins with 30% sequence similarity.Angiotensin I convert Angiotensin II human (Ang II) through removing of two C-terminal residues by the enzyme angiotensin-converting enzyme (ACE). purity: 99%
Catalog No. 50-257-6361 Supplier TARGETMOL CHEMICALS INC Supplier No. T704025MG
Add to Cart
Add to Cart
Also available in 5 mg 10 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Angiotensin II is mediated by AT1 and AT2 receptors. AT1 and AT2 receptors have seven transmembrane glycoproteins with 30% sequence similarity.Angiotensin I convert Angiotensin II human (Ang II) through removing of two C-terminal residues by the enzyme angiotensin-converting enzyme (ACE). purity: 99%