Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Angiotensin II, MP Biomedicals™

Catalog No. MP219505105 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219505105 5 mg
MP219505110 10 mg
MP219505101 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP219505105 Supplier MP Biomedicals, Inc Supplier No. 0219505105
Add to Cart
Add to Cart

Angiotensin II Human From Synthetic, Powder

  • Angiotensin II plays an important role in the regulation of blood pressure.
  • It stimulates the release of aldosterone from the adrenal gland.
  • Angiotensin II is a peptide hormone which plays an important role in the regulation of blood pressure.
  • It stimulates the release of aldosterone from the adrenal gland.It has strong vasoconstrictive effects.
TRUSTED_SUSTAINABILITY

Specifications

CAS 68521-88-0
Color Off-white
Content And Storage -20°C
Format Powder
Molecular Weight (g/mol) 1046
Quantity 5 mg
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.