Learn More
Bachem Angiotensin II Receptor Ligand
SDP
Supplier:  Bachem 4015751.0005
Encompass_Preferred
Angiotensin II Receptor Ligand, Nicotinoyl-Tyr-Lys(Z-Arg)-His-Pro-Ile-OH, 127060-75-7, C52H69N13O11, Mr 1052.20, 5mg. CGP42112A, CGP42112, Angiotensin AT2 Receptor Agonist. Depending on concentrations, CGP-42112 acts as a selective agonist or antagonist of AT2 receptor-mediated effects.
Catalog No. 50-260-240