Learn More
Enzo Life Sciences Angiotensin II receptor AT1 polyclonal antibody (100 µg)
SDP
Supplier:  Enzo Life Sciences ADI905743100
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to the sequence near the N-terminus of rat angiotensin II receptor AT1. UniProt: P25095 (AT1A), P29089 (AT1B). Species Reactivity: Human, Mouse, Rat. Crossreactivity: Predicted species reactivity based on epitope sequence homology: chimpanzee, bovine, rhesus macaque, sheep, dog, guinea pig, rabbit, horse, wild boar, gerbil. Applications: ELISA, IHC, WB. Purity: Peptide affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.01% sodium azide. Handling: Keep on ice at all times.
Catalog No. 50-658-497