Learn More
Medchemexpress LLC Angiotensin III, human, mouse | 13602-53-4 | 98.26% | 931.09 | 25 MG
SDP
Catalog No. 5000426610
Encompass_Preferred
Angiotensin III, human, mouse is a heptapeptide that functions as an endogenous angiotensin type 2 receptor (AT2R) agonist. It exhibits IC50s of 0.648 nM for AT2R and 21.1 nM for AT1R. This compound is intended for research use only.
  • Heptapeptide
  • Acts as an endogenous angiotensin type 2 receptor (AT2R) agonist
  • Exhibits IC50s of 0.648 nM for AT2R
  • Exhibits IC50s of 21.1 nM for AT1R
  • Increases urine sodium excretion in SD rats
  • Increases fractional excretion of sodium in SD rats
  • Increases fractional excretion of lithium in SD rats
Catalog No. 50-004-26610 Supplier Medchemexpress LLC Supplier No. 25MGHYP1540
Angiotensin III, human, mouse is a heptapeptide that functions as an endogenous angiotensin type 2 receptor (AT2R) agonist. It exhibits IC50s of 0.648 nM for AT2R and 21.1 nM for AT1R. This compound is intended for research use only.
  • Heptapeptide
  • Acts as an endogenous angiotensin type 2 receptor (AT2R) agonist
  • Exhibits IC50s of 0.648 nM for AT2R
  • Exhibits IC50s of 21.1 nM for AT1R
  • Increases urine sodium excretion in SD rats
  • Increases fractional excretion of sodium in SD rats
  • Increases fractional excretion of lithium in SD rats